Product Details

SNP ID
rs34777851
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:241108115 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGAGGAAATGGGAAAAAAAAGTG[G/T]CTGGTCTCTAAGGACAGTGTCGACT
Phenotype
MIM: 615393 MIM: 607505
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MTERF4 PubMed Links

Gene Details

Gene
MTERF4
Gene Name
mitochondrial transcription termination factor 4
There are no transcripts associated with this gene.

Gene
PASK
Gene Name
PAS domain containing serine/threonine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252119.1 Intron NP_001239048.1
NM_001252120.1 Intron NP_001239049.1
NM_001252122.1 Intron NP_001239051.1
NM_001252124.1 Intron NP_001239053.1
NM_015148.3 Intron NP_055963.2
XM_005246991.1 Intron XP_005247048.1
XM_011510826.1 Intron XP_011509128.1
XM_011510828.1 Intron XP_011509130.1
XM_011510829.1 Intron XP_011509131.1
XM_011510830.1 Intron XP_011509132.1
XM_011510831.2 Intron XP_011509133.1
XM_011510832.2 Intron XP_011509134.1
XM_011510833.1 Intron XP_011509135.1
XM_011510834.1 Intron XP_011509136.1
XM_011510835.1 Intron XP_011509137.1
XM_017003634.1 Intron XP_016859123.1
XM_017003635.1 Intron XP_016859124.1
XM_017003636.1 Intron XP_016859125.1

View Full Product Details