Product Details

SNP ID
rs4808867
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18877857 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTAACGCCGCATCTGCCAGAACCCA[C/T]GTGACCCTCTGCCCCAATCCCATCT
Phenotype
MIM: 606919 MIM: 602880 MIM: 601430
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CERS1 PubMed Links
Additional Information
For this assay, SNP(s) [rs200414993] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CERS1
Gene Name
ceramide synthase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290265.1 Intron NP_001277194.1
NM_021267.4 Intron NP_067090.1
NM_198207.2 Intron NP_937850.1
Gene
GDF1
Gene Name
growth differentiation factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001492.5 Intron NP_001483.3
Gene
UPF1
Gene Name
UPF1, RNA helicase and ATPase
There are no transcripts associated with this gene.

View Full Product Details