Product Details

SNP ID
rs35565092
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:100303781 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTTCACCCGCAGGCTCTGCAGCA[C/T]CAACCTGAGGACTCGAGGCAAGAGA
Phenotype
MIM: 243305 MIM: 616717
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
INVS PubMed Links

Gene Details

Gene
INVS
Gene Name
inversin
There are no transcripts associated with this gene.

Gene
TEX10
Gene Name
testis expressed 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161584.1 2231 Missense Mutation ATG,GTG M827V NP_001155056.1
NM_017746.3 2231 Missense Mutation ATG,GTG M843V NP_060216.2
XM_011518798.1 2231 Missense Mutation ATG,GTG M718V XP_011517100.1
XM_017014848.1 2231 Missense Mutation ATG,GTG M488V XP_016870337.1

View Full Product Details