Product Details

SNP ID
rs3782893
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:109855873 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCACCTACTGTGAGCCAGGAACATG[C/G]GCGCCCCAGAGGGAGTGGTTATTCC
Phenotype
MIM: 608949
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GLTP PubMed Links

Gene Details

Gene
GLTP
Gene Name
glycolipid transfer protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016433.3 Intron NP_057517.1
XM_011538434.2 Intron XP_011536736.1

View Full Product Details