Product Details

SNP ID
rs76066099
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:76739006 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGATATACCAGGTATTGTACCGT[A/G]TGATTGATTTTGCTTTATATTTGAC
Phenotype
MIM: 615262 MIM: 600813
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
METTL23 PubMed Links

Gene Details

Gene
METTL23
Gene Name
methyltransferase like 23
There are no transcripts associated with this gene.

Gene
MFSD11
Gene Name
major facilitator superfamily domain containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242532.1 Intron NP_001229461.1
NM_001242533.1 Intron NP_001229462.1
NM_001242534.1 Intron NP_001229463.1
NM_001242535.1 Intron NP_001229464.1
NM_001242536.1 Intron NP_001229465.1
NM_001242537.1 Intron NP_001229466.1
NM_024311.3 Intron NP_077287.1
XM_011525236.2 Intron XP_011523538.1
XM_011525237.2 Intron XP_011523539.2
XM_011525238.2 Intron XP_011523540.2
XM_011525239.2 Intron XP_011523541.2
XM_011525240.2 Intron XP_011523542.2
XM_011525241.2 Intron XP_011523543.2
XM_011525242.1 Intron XP_011523544.1
XM_011525244.1 Intron XP_011523546.1
XM_011525247.2 Intron XP_011523549.2
XM_017025065.1 Intron XP_016880554.1
XM_017025066.1 Intron XP_016880555.1
XM_017025067.1 Intron XP_016880556.1
XM_017025068.1 Intron XP_016880557.1
XM_017025069.1 Intron XP_016880558.1
XM_017025070.1 Intron XP_016880559.1
XM_017025071.1 Intron XP_016880560.1
Gene
MIR636
Gene Name
microRNA 636
There are no transcripts associated with this gene.

Gene
SRSF2
Gene Name
serine and arginine rich splicing factor 2
There are no transcripts associated with this gene.

View Full Product Details