Product Details

SNP ID
rs3209157
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:72223837 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAAGTCAGAGCCTTCAGCACTGCA[C/T]CCAGCACATCGAGCTGTGGAGCAGA
Phenotype
MIM: 611989
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MRPS27 PubMed Links

Gene Details

Gene
MRPS27
Gene Name
mitochondrial ribosomal protein S27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286748.1 855 Missense Mutation GAT,GGT D298G NP_001273677.1
NM_001286751.1 855 Missense Mutation GAT,GGT D228G NP_001273680.1
NM_015084.2 855 Missense Mutation GAT,GGT D284G NP_055899.2

View Full Product Details