Product Details

SNP ID
rs33998366
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:69180296 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGTAGAACCTGCGGCCTCGATGT[C/T]CTTCTCCCGTGCCCTATTGTGGGCT
Phenotype
MIM: 605122 MIM: 605506
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SUPV3L1 PubMed Links

Gene Details

Gene
SUPV3L1
Gene Name
Suv3 like RNA helicase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301683.1 93 UTR 5 NP_001288612.1
NM_001323584.1 93 UTR 5 NP_001310513.1
NM_001323585.1 93 UTR 5 NP_001310514.1
NM_001323586.1 93 UTR 5 NP_001310515.1
NM_001323587.1 93 UTR 5 NP_001310516.1
NM_001323588.1 93 UTR 5 NP_001310517.1
NM_003171.4 93 Missense Mutation TCC,TTC S2F NP_003162.2
Gene
VPS26A
Gene Name
VPS26, retromer complex component A
There are no transcripts associated with this gene.

View Full Product Details