Product Details

SNP ID
rs7133238
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6451931 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGTTATAGCTCCACTTTTTTGGGA[A/C]AGGTGGGGAGCAAAAGGGTGCAGAG
Phenotype
MIM: 186711 MIM: 607081
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CD27 PubMed Links

Gene Details

Gene
CD27
Gene Name
CD27 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242.4 Intron NP_001233.1
XM_011521042.2 Intron XP_011519344.1
XM_017020232.1 Intron XP_016875721.1
XM_017020233.1 Intron XP_016875722.1
XM_017020234.1 Intron XP_016875723.1
Gene
CD27-AS1
Gene Name
CD27 antisense RNA 1
There are no transcripts associated with this gene.

Gene
TAPBPL
Gene Name
TAP binding protein like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018009.4 Intron NP_060479.3
XM_005253700.3 Intron XP_005253757.1
XM_017019545.1 Intron XP_016875034.1
XM_017019546.1 Intron XP_016875035.1

View Full Product Details