Product Details

SNP ID
rs6675153
Assay Type
Functionally tested
NCBI dbSNP Submissions
49
Location
Chr.1:166857536 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAATTTTCAGTAATCAATGAATT[C/T]GGTGAGGGTCCCACACCCTCAAATC
Phenotype
MIM: 612763
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POGK PubMed Links

Gene Details

Gene
POGK
Gene Name
pogo transposable element with KRAB domain
There are no transcripts associated with this gene.

Gene
TADA1
Gene Name
transcriptional adaptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_053053.3 Intron NP_444281.1

View Full Product Details