Product Details

SNP ID
rs35492126
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45793987 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGCAGCCCAGCATCAACGAGGA[A/G]ATCCAGACTGTCTTCAACAAGTACA
Phenotype
MIM: 611388
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNTTIP1 PubMed Links

Gene Details

Gene
DNTTIP1
Gene Name
deoxynucleotidyltransferase, terminal, interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052951.2 311 Silent Mutation GAA,GAG E81E NP_443183.1
Gene
WFDC3
Gene Name
WAP four-disulfide core domain 3
There are no transcripts associated with this gene.

View Full Product Details