Product Details

SNP ID
rs17197261
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:21570031 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTACATGGGCTCCACAAGTTGAAAA[A/C]GCCCGGCGCCGCCCATCAGCTGTGT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR10G3 PubMed Links

Gene Details

Gene
OR10G3
Gene Name
olfactory receptor family 10 subfamily G member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005465.1 714 Silent Mutation GCG,GCT A238A NP_001005465.1

View Full Product Details