Product Details
- SNP ID
-
rs11000226
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:72223766 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AATAAAATATTGTGAAAGTAGAGAG[C/G]ATAGAAAAAAGGGGAGCTAACTCTT
- Phenotype
-
MIM: 613427
MIM: 614215
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ANAPC16
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs146378629] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ANAPC16
- Gene Name
- anaphase promoting complex subunit 16
- Gene
- ASCC1
- Gene Name
- activating signal cointegrator 1 complex subunit 1
There are no transcripts associated with this gene.
View Full Product Details