Product Details

SNP ID
rs200534634
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:4461742 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGTTGGTGCCTCTGCCCCTCTGG[A/T]GCCCCCGATCCCCACAAGGGGCCGC
Phenotype
MIM: 608382
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DNAJA3 PubMed Links

Gene Details

Gene
DNAJA3
Gene Name
DnaJ heat shock protein family (Hsp40) member A3
There are no transcripts associated with this gene.

Gene
NMRAL1
Gene Name
NmrA-like family domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305141.1 1251 UTR 3 NP_001292070.1
NM_001305142.1 1251 UTR 3 NP_001292071.1
NM_020677.4 1251 UTR 3 NP_065728.1
XM_006720905.3 1251 UTR 3 XP_006720968.1
XM_006720906.2 1251 UTR 3 XP_006720969.1
XM_011522567.2 1251 Intron XP_011520869.1
XM_017023485.1 1251 UTR 3 XP_016878974.1
XM_017023486.1 1251 Intron XP_016878975.1
XM_017023487.1 1251 Intron XP_016878976.1
XM_017023488.1 1251 Intron XP_016878977.1
XM_017023489.1 1251 UTR 3 XP_016878978.1
XM_017023490.1 1251 UTR 3 XP_016878979.1
XM_017023491.1 1251 UTR 3 XP_016878980.1
XM_017023492.1 1251 UTR 3 XP_016878981.1

View Full Product Details