Product Details

SNP ID
rs201308166
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:53797481 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGTAATTCAGCTGAGTTATGTGG[C/G]TTTTGTGCATTGAAATATATCCATC
Phenotype
MIM: 611229
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ERLEC1 PubMed Links

Gene Details

Gene
ERLEC1
Gene Name
endoplasmic reticulum lectin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127397.2 Intron NP_001120869.1
NM_001127398.2 Intron NP_001120870.1
NM_015701.4 Intron NP_056516.2
XM_011532766.1 Intron XP_011531068.1
Gene
GPR75-ASB3
Gene Name
GPR75-ASB3 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164165.1 Intron NP_001157637.1

View Full Product Details