Product Details

SNP ID
rs61754909
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:73291659 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCATTTACTGACTTCAGATTGGGG[G/T]ACCTGAAGTGAAAAAGCAAACATAA
Phenotype
MIM: 607599 MIM: 604166
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C8orf89 PubMed Links

Gene Details

Gene
C8orf89
Gene Name
chromosome 8 open reading frame 89
There are no transcripts associated with this gene.

Gene
RDH10
Gene Name
retinol dehydrogenase 10 (all-trans)
There are no transcripts associated with this gene.

Gene
RPL7
Gene Name
ribosomal protein L7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000971.3 1046 Intron NP_000962.2
XM_006716463.2 1046 Missense Mutation TAC,TCC Y104S XP_006716526.1

View Full Product Details