Product Details

SNP ID
rs3738863
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:30234695 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGCAACAATCGTCCTTATCTCCTA[A/G]GATTGTGGTAAAGATTCACCATGAC
Phenotype
MIM: 611763
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LBH PubMed Links

Gene Details

Gene
LBH
Gene Name
limb bud and heart development
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030915.3 Intron NP_112177.2
XM_017005059.1 Intron XP_016860548.1

View Full Product Details