Product Details

SNP ID
rs61754950
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:133120359 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTCTGACTGAAGGCTTTTCCACA[C/T]TGAATACATTCATAGGGTTTCTCTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF891 PubMed Links

Gene Details

Gene
ZNF891
Gene Name
zinc finger protein 891
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277291.1 1751 Silent Mutation CAA,CAG Q520Q NP_001264220.1
XM_017018666.1 1751 Silent Mutation CAA,CAG Q520Q XP_016874155.1
XM_017018667.1 1751 Silent Mutation CAA,CAG Q520Q XP_016874156.1
XM_017018668.1 1751 Silent Mutation CAA,CAG Q520Q XP_016874157.1

View Full Product Details