Product Details

SNP ID
rs35805933
Assay Type
Functionally tested
NCBI dbSNP Submissions
27
Location
Chr.1:36393977 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTAGGTGACTTTGGCCAGGCGGAT[A/G]TTCATGAAAGCATCGACATTGTCTA
Phenotype
MIM: 609437
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LSM10 PubMed Links

Gene Details

Gene
LSM10
Gene Name
LSM10, U7 small nuclear RNA associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032881.2 370 Silent Mutation AAC,AAT N51N NP_116270.1
Gene
STK40
Gene Name
serine/threonine kinase 40
There are no transcripts associated with this gene.

View Full Product Details