Product Details

SNP ID
rs116276069
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:36496490 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACCCCACAGAGCAGACGCTAAAA[A/G]TGCAAGAAGGTATGGACAAGTACTA
Phenotype
MIM: 604500
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MYO19 PubMed Links

Gene Details

Gene
MYO19
Gene Name
myosin XIX
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033580.2 Intron NP_001028752.1
NM_001163735.1 Intron NP_001157207.1
NM_025109.5 Intron NP_079385.2
XM_011525286.1 Intron XP_011523588.1
XM_011525287.1 Intron XP_011523589.1
XM_011525290.2 Intron XP_011523592.1
XM_017025157.1 Intron XP_016880646.1
XM_017025158.1 Intron XP_016880647.1
XM_017025159.1 Intron XP_016880648.1
XM_017025160.1 Intron XP_016880649.1
XM_017025161.1 Intron XP_016880650.1
Gene
ZNHIT3
Gene Name
zinc finger HIT-type containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281432.1 Intron NP_001268361.1
NM_001281433.1 Intron NP_001268362.1
NM_001281434.1 Intron NP_001268363.1
NM_004773.3 Intron NP_004764.1

View Full Product Details