Product Details

SNP ID
hCV25987387
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:119469989 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTCTAACATCCTTGAGAGATTTT[A/G]GTTGCAGTGGTCAGAAAGGCAAGGA
Phenotype
MIM: 615618
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
POGLUT1 PubMed Links

Gene Details

Gene
POGLUT1
Gene Name
protein O-glucosyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152305.2 Intron NP_689518.1
Gene
TMEM39A
Gene Name
transmembrane protein 39A
There are no transcripts associated with this gene.

View Full Product Details