Product Details

SNP ID
hCV25987584
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:41028489 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCTGTTCTCCTCAGCCTTTGTCT[A/C]TAAGCATCCCTCCTCCCCAAACATC
Phenotype
MIM: 616396
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SCMH1 PubMed Links

Gene Details

Gene
SCMH1
Gene Name
sex comb on midleg homolog 1 (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031694.2 Intron NP_001026864.1
NM_001172218.1 Intron NP_001165689.1
NM_001172219.1 Intron NP_001165690.1
NM_001172220.1 Intron NP_001165691.1
NM_001172221.1 Intron NP_001165692.1
NM_001172222.2 Intron NP_001165693.1
NM_012236.3 Intron NP_036368.1
Gene
SLFNL1
Gene Name
schlafen like 1
There are no transcripts associated with this gene.

Gene
SLFNL1-AS1
Gene Name
SLFNL1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details