Product Details

SNP ID
rs10769224
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5788443 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACGCAGAGGGAAGCAGATGGCCACA[C/T]AGTGGTCCAGGGCCATGAGCATGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR52N1 PubMed Links

Gene Details

Gene
OR52N1
Gene Name
olfactory receptor family 52 subfamily N member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001913.1 374 Missense Mutation TAT,TGT Y125C NP_001001913.1
Gene
OR52N5
Gene Name
olfactory receptor family 52 subfamily N member 5
There are no transcripts associated with this gene.

View Full Product Details