Product Details

SNP ID
rs113010274
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:113446583 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCATTTCACAATTACATTTTAAC[A/G]TGTACCTTTGCTCTTGGATTAAGAG
Phenotype
MIM: 613447
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CFAP44 PubMed Links

Gene Details

Gene
CFAP44
Gene Name
cilia and flagella associated protein 44
There are no transcripts associated with this gene.

Gene
SPICE1
Gene Name
spindle and centriole associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144718.3 Intron NP_653319.1
XM_011512461.2 Intron XP_011510763.1
XM_017005775.1 Intron XP_016861264.1
XM_017005776.1 Intron XP_016861265.1
XM_017005777.1 Intron XP_016861266.1
XM_017005778.1 Intron XP_016861267.1

View Full Product Details