Product Details

SNP ID
rs9934889
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:19410697 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGAAGGCCGGCTCCGGTGCAGGG[A/C]GCGCCGCCTGCGGGCTGCTTCGGGC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TMC5 PubMed Links

Gene Details

Gene
TMC5
Gene Name
transmembrane channel like 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105248.1 159 Intron NP_001098718.1
NM_001105249.1 159 Intron NP_001098719.1
NM_001261841.1 159 Intron NP_001248770.1
NM_001308161.1 159 UTR 5 NP_001295090.1
NM_024780.4 159 Intron NP_079056.2

View Full Product Details