Product Details

SNP ID
rs61742446
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:149765553 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGTCTGGTGCACCCTTTGGTGCC[G/T]CACCAAGTGCTGCTTGTGCGTGAAG
Phenotype
MIM: 614040
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF467 PubMed Links

Gene Details

Gene
ZNF467
Gene Name
zinc finger protein 467
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005249959.4 1062 Silent Mutation AGG,CGG R317R XP_005250016.1
XM_005249960.4 1062 Silent Mutation AGG,CGG R317R XP_005250017.1
XM_005249961.4 1062 Silent Mutation AGG,CGG R317R XP_005250018.1
XM_006715864.3 1062 Silent Mutation AGG,CGG R358R XP_006715927.1
XM_011515855.2 1062 Silent Mutation AGG,CGG R568R XP_011514157.1
XM_011515856.2 1062 Silent Mutation AGG,CGG R568R XP_011514158.1
XM_011515857.2 1062 Silent Mutation AGG,CGG R531R XP_011514159.1
XM_011515858.1 1062 Silent Mutation AGG,CGG R317R XP_011514160.1
XM_017011799.1 1062 Silent Mutation AGG,CGG R492R XP_016867288.1

View Full Product Details