Product Details

SNP ID
rs17040904
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:109851555 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAACGAGCTGCCTACGAACCTCCC[C/T]GTGGACACTGTGAAGCTTCGCATAG
Phenotype
MIM: 615004 MIM: 605224
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRIT3 PubMed Links

Gene Details

Gene
LRIT3
Gene Name
leucine rich repeat, Ig-like and transmembrane domains 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198506.4 188 Silent Mutation CCC,CCT P56P NP_940908.3
XM_017008167.1 188 Intron XP_016863656.1
XM_017008168.1 188 Silent Mutation CCC,CCT P56P XP_016863657.1
Gene
RRH
Gene Name
retinal pigment epithelium-derived rhodopsin homolog
There are no transcripts associated with this gene.

View Full Product Details