Product Details

SNP ID
hCV25990649
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2271704 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGAGCTCAACACGGCCTAGGCC[C/G]AAGCTTTAGCGCTGCCGGGTTCTTG
Phenotype
MIM: 601579
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C19orf35 PubMed Links

Gene Details

Gene
C19orf35
Gene Name
chromosome 19 open reading frame 35
There are no transcripts associated with this gene.

Gene
OAZ1
Gene Name
ornithine decarboxylase antizyme 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301020.1 Intron NP_001287949.1
NM_004152.3 Intron NP_004143.1

View Full Product Details