Product Details

SNP ID
rs564473772
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:108874643 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTGCCTTTATGAACAAGCATATG[C/T]CACATAAATATTTTTAAAAATTACA
Phenotype
MIM: 603722
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IKBKAP PubMed Links

Gene Details

Gene
IKBKAP
Gene Name
inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase complex-associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318360.1 Intron NP_001305289.1
NM_003640.4 Intron NP_003631.2
XM_011519136.1 Intron XP_011517438.1
XM_011519137.1 Intron XP_011517439.1
XM_017015238.1 Intron XP_016870727.1

View Full Product Details