Product Details
- SNP ID
-
rs34492126
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:197044642 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGTTTTCATAGCTTTTCTTTTGCC[G/A]GAACCCAGATGTAAGAACCAGATTG
- Phenotype
-
MIM: 601014
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DLG1
PubMed Links
Gene Details
- Gene
- DLG1
- Gene Name
- discs large MAGUK scaffold protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001098424.1 |
2839 |
Missense Mutation |
CCG,CTG |
P899L |
NP_001091894.1 |
NM_001204386.1 |
2839 |
Missense Mutation |
CCG,CTG |
P887L |
NP_001191315.1 |
NM_001204387.1 |
2839 |
Missense Mutation |
CCG,CTG |
P795L |
NP_001191316.1 |
NM_001204388.1 |
2839 |
Missense Mutation |
CCG,CTG |
P783L |
NP_001191317.1 |
NM_001290983.1 |
2839 |
Missense Mutation |
CCG,CTG |
P899L |
NP_001277912.1 |
NM_004087.2 |
2839 |
Missense Mutation |
CCG,CTG |
P921L |
NP_004078.2 |
XM_005269289.3 |
2839 |
Missense Mutation |
CCG,CTG |
P921L |
XP_005269346.1 |
XM_011512502.2 |
2839 |
Missense Mutation |
CCG,CTG |
P899L |
XP_011510804.1 |
XM_011512503.1 |
2839 |
Missense Mutation |
CCG,CTG |
P887L |
XP_011510805.1 |
XM_011512505.1 |
2839 |
Missense Mutation |
CCG,CTG |
P866L |
XP_011510807.1 |
XM_011512506.1 |
2839 |
Missense Mutation |
CCG,CTG |
P848L |
XP_011510808.1 |
XM_011512509.1 |
2839 |
Intron |
|
|
XP_011510811.1 |
XM_017005800.1 |
2839 |
Missense Mutation |
CCG,CTG |
P921L |
XP_016861289.1 |
XM_017005801.1 |
2839 |
Missense Mutation |
CCG,CTG |
P921L |
XP_016861290.1 |
XM_017005802.1 |
2839 |
Missense Mutation |
CCG,CTG |
P921L |
XP_016861291.1 |
XM_017005803.1 |
2839 |
Missense Mutation |
CCG,CTG |
P921L |
XP_016861292.1 |
XM_017005804.1 |
2839 |
Missense Mutation |
CCG,CTG |
P920L |
XP_016861293.1 |
XM_017005805.1 |
2839 |
Missense Mutation |
CCG,CTG |
P899L |
XP_016861294.1 |
XM_017005806.1 |
2839 |
Missense Mutation |
CCG,CTG |
P888L |
XP_016861295.1 |
XM_017005807.1 |
2839 |
Missense Mutation |
CCG,CTG |
P888L |
XP_016861296.1 |
XM_017005808.1 |
2839 |
Missense Mutation |
CCG,CTG |
P888L |
XP_016861297.1 |
XM_017005809.1 |
2839 |
Missense Mutation |
CCG,CTG |
P888L |
XP_016861298.1 |
XM_017005810.1 |
2839 |
Missense Mutation |
CCG,CTG |
P887L |
XP_016861299.1 |
XM_017005811.1 |
2839 |
Missense Mutation |
CCG,CTG |
P870L |
XP_016861300.1 |
XM_017005812.1 |
2839 |
Missense Mutation |
CCG,CTG |
P870L |
XP_016861301.1 |
XM_017005813.1 |
2839 |
Missense Mutation |
CCG,CTG |
P870L |
XP_016861302.1 |
XM_017005814.1 |
2839 |
Missense Mutation |
CCG,CTG |
P870L |
XP_016861303.1 |
XM_017005815.1 |
2839 |
Missense Mutation |
CCG,CTG |
P869L |
XP_016861304.1 |
XM_017005816.1 |
2839 |
Missense Mutation |
CCG,CTG |
P866L |
XP_016861305.1 |
XM_017005817.1 |
2839 |
Missense Mutation |
CCG,CTG |
P866L |
XP_016861306.1 |
XM_017005818.1 |
2839 |
Missense Mutation |
CCG,CTG |
P866L |
XP_016861307.1 |
XM_017005819.1 |
2839 |
Missense Mutation |
CCG,CTG |
P866L |
XP_016861308.1 |
XM_017005820.1 |
2839 |
Missense Mutation |
CCG,CTG |
P848L |
XP_016861309.1 |
XM_017005821.1 |
2839 |
Missense Mutation |
CCG,CTG |
P838L |
XP_016861310.1 |
XM_017005822.1 |
2839 |
Missense Mutation |
CCG,CTG |
P816L |
XP_016861311.1 |
XM_017005823.1 |
2839 |
Missense Mutation |
CCG,CTG |
P805L |
XP_016861312.1 |
XM_017005824.1 |
2839 |
Missense Mutation |
CCG,CTG |
P514L |
XP_016861313.1 |
View Full Product Details