Product Details

SNP ID
rs61745624
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30103785 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTTGGTGGCCCCGATGTAGTTCT[G/T]CCGCCTTTGCGGGAGAAGGAAAGGA
Phenotype
MIM: 609316
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TRIM31 PubMed Links

Gene Details

Gene
TRIM31
Gene Name
tripartite motif containing 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007028.4 1139 Silent Mutation GGA,GGC G343G NP_008959.3
XM_011514264.1 1139 Silent Mutation GGA,GGC G342G XP_011512566.1
XM_011514265.1 1139 Silent Mutation GGA,GGC G343G XP_011512567.1
XM_011514266.1 1139 Intron XP_011512568.1
Gene
TRIM31-AS1
Gene Name
TRIM31 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details