Product Details

SNP ID
rs375930905
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:38255024 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTTACTGAATTGTCTTGTTCAGG[A/G]ACAAGTGGTGTCTCTCCCAGCTTCT
Phenotype
MIM: 616845
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLEC14A PubMed Links

Gene Details

Gene
CLEC14A
Gene Name
C-type lectin domain family 14 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175060.2 1347 Silent Mutation GTC,GTT V333V NP_778230.1

View Full Product Details