Product Details

SNP ID
rs61739527
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:52156619 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATATATCACATTGATATGGTTTC[C/T]CTCCTGTATGGATTATCTGATGTGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF836 PubMed Links

Gene Details

Gene
ZNF836
Gene Name
zinc finger protein 836
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102657.1 1473 Missense Mutation GAG,GGG E355G NP_001096127.1
XM_011526558.2 1473 Missense Mutation GAG,GGG E355G XP_011524860.1
XM_011526559.2 1473 Missense Mutation GAG,GGG E355G XP_011524861.1
XM_017026389.1 1473 Intron XP_016881878.1

View Full Product Details