Product Details

SNP ID
rs3743567
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:58516580 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAGCGAGTACCAGTCCATCGTGCT[A/G]CCCTTCATGGAAGCCCACCCCGATC
Phenotype
MIM: 604917 MIM: 614463 MIM: 616424
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNOT1 PubMed Links

Gene Details

Gene
CNOT1
Gene Name
CCR4-NOT transcription complex subunit 1
There are no transcripts associated with this gene.

Gene
NDRG4
Gene Name
NDRG family member 4
There are no transcripts associated with this gene.

Gene
SETD6
Gene Name
SET domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160305.2 566 Silent Mutation CTA,CTG L193L NP_001153777.1
NM_024860.3 566 Silent Mutation CTA,CTG L169L NP_079136.2
XM_011523336.1 566 Silent Mutation CTA,CTG L193L XP_011521638.1

View Full Product Details