Product Details

SNP ID
rs34982573
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:82827568 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTTGCTGCAGCATTTGTGACTGG[A/G]GGTCACCCAACGGGTTCATGATTGG
Phenotype
MIM: 610257
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SEC31A PubMed Links

Gene Details

Gene
SEC31A
Gene Name
SEC31 homolog A, COPII coat complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077206.3 3172 Missense Mutation NP_001070674.1
NM_001077207.3 3172 Missense Mutation NP_001070675.1
NM_001077208.3 3172 Missense Mutation NP_001070676.1
NM_001191049.2 3172 Missense Mutation NP_001177978.1
NM_001300744.2 3172 Missense Mutation NP_001287673.1
NM_001300745.2 3172 Missense Mutation NP_001287674.1
NM_001318119.1 3172 Missense Mutation NP_001305048.1
NM_001318120.1 3172 Missense Mutation NP_001305049.1
NM_016211.4 3172 Missense Mutation NP_057295.2

View Full Product Details