Product Details

SNP ID
rs190680760
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:76867504 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTTTTAATTCTGTCTCTAATCTC[C/T]GCAACAGCCGCGCTTCCCGGGTCCC
Phenotype
MIM: 610134
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ST6GALNAC5 PubMed Links

Gene Details

Gene
ST6GALNAC5
Gene Name
ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320273.1 97 UTR 5 NP_001307202.1
NM_001320274.1 97 UTR 5 NP_001307203.1
NM_030965.2 97 UTR 5 NP_112227.1

View Full Product Details