Product Details

SNP ID
rs138304742
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:77092549 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGGTAAATAATTCAGCAATCAAT[A/G]AAGCAAACATTGATGAAAAAGATAA
Phenotype
MIM: 605087
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PIGK PubMed Links

Gene Details

Gene
PIGK
Gene Name
phosphatidylinositol glycan anchor biosynthesis class K
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005482.2 Intron NP_005473.1

View Full Product Details