Product Details
- SNP ID
-
rs114064150
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:85433130 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGCATCGTCCAGAGTGGTGAGCAG[C/T]CCCTCCATTTCCACCTCAGTTTCCA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SH2D6
PubMed Links
Gene Details
- Gene
- SH2D6
- Gene Name
- SH2 domain containing 6
View Full Product Details