Product Details

SNP ID
rs113387655
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1358264 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGATGAATTCCTAGTTGGCTGT[A/G]ACTTTTGGTTTAAGTGGAAGGTTGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MUM1 PubMed Links

Gene Details

Gene
MUM1
Gene Name
melanoma associated antigen (mutated) 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032853.3 Intron NP_116242.2
XM_011528380.2 Intron XP_011526682.1
XM_011528382.2 Intron XP_011526684.1
XM_011528383.1 Intron XP_011526685.1
XM_011528384.2 Intron XP_011526686.1
XM_011528385.2 Intron XP_011526687.1
XM_017027401.1 Intron XP_016882890.1

View Full Product Details