Product Details

SNP ID
rs543355515
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63241638 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGACGGCTTCAGGCCTTTGGCA[A/G]AAAGGAGATGGGTGGGACTGAGGCT
Phenotype
MIM: 605737 MIM: 612873
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BIRC7 PubMed Links

Gene Details

Gene
BIRC7
Gene Name
baculoviral IAP repeat containing 7
There are no transcripts associated with this gene.

Gene
LOC107985425
Gene Name
uncharacterized LOC107985425
There are no transcripts associated with this gene.

Gene
MIR3196
Gene Name
microRNA 3196
There are no transcripts associated with this gene.

Gene
NKAIN4
Gene Name
Na+/K+ transporting ATPase interacting 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152864.3 1236 Intron NP_690603.3
XM_005260192.1 1236 Intron XP_005260249.1
XM_005260194.3 1236 Intron XP_005260251.1
XM_011528527.1 1236 UTR 3 XP_011526829.1
XM_011528528.2 1236 UTR 3 XP_011526830.1
XM_011528529.2 1236 UTR 3 XP_011526831.1
XM_017027636.1 1236 UTR 3 XP_016883125.1
XM_017027637.1 1236 UTR 3 XP_016883126.1
XM_017027638.1 1236 Intron XP_016883127.1
XM_017027639.1 1236 Intron XP_016883128.1

View Full Product Details