Product Details

SNP ID
rs145834782
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:19896986 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAATACTTCCATTAAGTTAAAAAT[A/G]TCAATAATTTTTTGCAACATTTCTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TTC32 PubMed Links

Gene Details

Gene
TTC32
Gene Name
tetratricopeptide repeat domain 32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008237.2 589 UTR 3 NP_001008238.1

View Full Product Details