Product Details

SNP ID
rs369805758
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:37495508 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCCCCACCCACTACCTGCCCAGC[C/T]GTGCTCACATTACTGCGGGGGCCTC
Phenotype
MIM: 607209 MIM: 602577
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CARD10 PubMed Links

Gene Details

Gene
CARD10
Gene Name
caspase recruitment domain family member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014550.3 Intron NP_055365.2
Gene
MFNG
Gene Name
MFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase
There are no transcripts associated with this gene.

View Full Product Details