Product Details

SNP ID
rs7540491
Assay Type
Functionally tested
NCBI dbSNP Submissions
64
Location
Chr.1:24057329 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACAGGTTGTCCCTACTGGTCCATG[C/T]AGACTAGACTCAGACTGTGCCTGGA
Phenotype
MIM: 616832
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYOM3 PubMed Links

Gene Details

Gene
MYOM3
Gene Name
myomesin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152372.3 4512 UTR 3 NP_689585.3

View Full Product Details