Product Details

SNP ID
rs4141491
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:46709763 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTTGAGTAGGAAGCCTGAGTCAGT[A/G]GAACAGATTAATCATTTAGGCAGAT
Phenotype
MIM: 612147
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MYLK3 PubMed Links

Gene Details

Gene
MYLK3
Gene Name
myosin light chain kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308301.1 Intron NP_001295230.1
NM_182493.2 Intron NP_872299.2
XM_006721334.3 Intron XP_006721397.1

View Full Product Details