Product Details

SNP ID
rs61739476
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:128872628 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTCTCCCACCCTTGGTCTGCACC[A/G]CTCCCGGTTTCACCCAGATCTGGGG
Phenotype
MIM: 607160 MIM: 609344
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP6V1F PubMed Links

Gene Details

Gene
ATP6V1F
Gene Name
ATPase H+ transporting V1 subunit F
There are no transcripts associated with this gene.

Gene
KCP
Gene Name
kielin/chordin-like protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135914.1 4721 Intron NP_001129386.1
NM_199349.2 4721 Intron NP_955381.2
XM_017012184.1 4721 Intron XP_016867673.1
XM_017012185.1 4721 Intron XP_016867674.1
XM_017012186.1 4721 UTR 3 XP_016867675.1
XM_017012187.1 4721 Intron XP_016867676.1
XM_017012188.1 4721 UTR 3 XP_016867677.1
XM_017012189.1 4721 UTR 3 XP_016867678.1
XM_017012190.1 4721 Intron XP_016867679.1
XM_017012191.1 4721 Intron XP_016867680.1
XM_017012192.1 4721 Intron XP_016867681.1
XM_017012193.1 4721 Intron XP_016867682.1
XM_017012194.1 4721 Intron XP_016867683.1
Gene
LOC100130705
Gene Name
uncharacterized LOC100130705
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195150.1 4721 Intron NP_001182079.1

View Full Product Details