Product Details

SNP ID
rs573520802
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:44989585 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCCATTCTTCGCTGCTGATCGC[G/T]GGATTCTTTTTGGATAGGGTTGACG
Phenotype
MIM: 604354
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GPALPP1 PubMed Links

Gene Details

Gene
GPALPP1
Gene Name
GPALPP motifs containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316951.1 56 UTR 5 NP_001303880.1
NM_001316952.1 56 UTR 5 NP_001303881.1
NM_018559.3 56 UTR 5 NP_061029.2
XM_005266444.3 56 UTR 5 XP_005266501.1
XM_011535144.2 56 Intron XP_011533446.1
Gene
NUFIP1
Gene Name
NUFIP1, FMR1 interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012345.2 56 Intron NP_036477.2

View Full Product Details