Product Details

SNP ID
rs61741960
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:44504510 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTGCTAGCTTAGGTACAACCTGA[C/T]TACAGTTCAGGAGGACGTTGTTTTA
Phenotype
MIM: 611779
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FSCB PubMed Links

Gene Details

Gene
FSCB
Gene Name
fibrous sheath CABYR binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032135.3 2787 Silent Mutation TAA,TAG *826* NP_115511.3
Gene
LOC105370473
Gene Name
uncharacterized LOC105370473
There are no transcripts associated with this gene.

View Full Product Details