Product Details

SNP ID
rs17115207
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:44505548 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGAGTTTCATCTGCAGGAGGCTC[C/T]GTAGCTGCTAGAAGCTGAATTTCAG
Phenotype
MIM: 611779
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FSCB PubMed Links

Gene Details

Gene
FSCB
Gene Name
fibrous sheath CABYR binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032135.3 1749 Silent Mutation ACA,ACG T480T NP_115511.3
Gene
LOC105370473
Gene Name
uncharacterized LOC105370473
There are no transcripts associated with this gene.

View Full Product Details