Product Details
- SNP ID
-
hCV25997721
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:91969965 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAAAATGAAATAACTCTGAATGTA[A/G]TAGCATAAAGTTATCTGTGTAATAT
- Phenotype
-
MIM: 604505
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TRIP11
PubMed Links
Gene Details
- Gene
- TRIP11
- Gene Name
- thyroid hormone receptor interactor 11
There are no transcripts associated with this gene.
View Full Product Details