Product Details

SNP ID
rs80052372
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67976074 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCTCAAACCACTGAACCATCCT[G/T]TCCACCAGCCCAGCTTACCTTTTCC
Phenotype
MIM: 609926 MIM: 604119
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DPEP3 PubMed Links

Gene Details

Gene
DPEP3
Gene Name
dipeptidase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001129758.1 Intron NP_001123230.1
NM_022357.3 Intron NP_071752.3
Gene
SLC12A4
Gene Name
solute carrier family 12 member 4
There are no transcripts associated with this gene.

View Full Product Details