Product Details

SNP ID
rs16967754
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75263020 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGTGTACGGGCTTTAATGAAGCCC[C/T]GTAAGAGTCAGCAACTGCCATTCCT
Phenotype
MIM: 606006 MIM: 612072 MIM: 611974 MIM: 606521
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
GGA3 PubMed Links
Additional Information
For this assay, SNP(s) [rs115214743] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
GGA3
Gene Name
golgi associated, gamma adaptin ear containing, ARF binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172703.2 Intron NP_001166174.1
NM_001172704.2 Intron NP_001166175.1
NM_001291641.1 Intron NP_001278570.1
NM_001291642.1 Intron NP_001278571.1
NM_014001.4 Intron NP_054720.1
NM_138619.3 Intron NP_619525.1
XM_011524563.2 Intron XP_011522865.1
XM_017024385.1 Intron XP_016879874.1
XM_017024386.1 Intron XP_016879875.1
XM_017024387.1 Intron XP_016879876.1
XM_017024388.1 Intron XP_016879877.1
XM_017024389.1 Intron XP_016879878.1
XM_017024390.1 Intron XP_016879879.1
XM_017024391.1 Intron XP_016879880.1
XM_017024392.1 Intron XP_016879881.1
Gene
LOC100287042
Gene Name
uncharacterized LOC100287042
There are no transcripts associated with this gene.

Gene
MIF4GD
Gene Name
MIF4G domain containing
There are no transcripts associated with this gene.

Gene
MRPS7
Gene Name
mitochondrial ribosomal protein S7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015971.3 Intron NP_057055.2
Gene
SLC25A19
Gene Name
solute carrier family 25 member 19
There are no transcripts associated with this gene.

View Full Product Details